Learn More
Cayman Chemical ClarIthromycIn-13C-d3 500ug
SDP
Catalog No. 502934525
Encompass_Preferred
An internal standard for the quantification of clarithromycin by GC- or LC-MS
Catalog No. 50-293-4525 Supplier Cayman Chemical Supplier No. 26678500 UG
An internal standard for the quantification of clarithromycin by GC- or LC-MS