Learn More
Cayman Chemical ClarIthromycIn 1g
SDP
Catalog No. 502981929
Encompass_Preferred
A macrolide antibiotic; active against methicillin-susceptible, but not methicillin-resistant, S. aureus (MIC50s = 0.06 and >128 UG/ml), S. pyogenes, L. monocytogenes, and B. pertussis (MIC50 = 0.015, 0.25, and ≤0.008 UG/ml, respectively), among others; decreases the number of CFUs in the spleen in a mouse model of M. avium infection
Catalog No. 50-298-1929 Supplier Cayman Chemical Supplier No. 194551 G
A macrolide antibiotic; active against methicillin-susceptible, but not methicillin-resistant, S. aureus (MIC50s = 0.06 and >128 UG/ml), S. pyogenes, L. monocytogenes, and B. pertussis (MIC50 = 0.015, 0.25, and ≤0.008 UG/ml, respectively), among others; decreases the number of CFUs in the spleen in a mouse model of M. avium infection