Learn More
Selleck Chemical LLC Clarithromycin S2555-1g
SDP
Catalog No. 503112936
Encompass_Preferred
Clarithromycin (A-56268) is a macrolide antibiotic and a CYP3A4 inhibitor used to treat a number of bacterial infections
Catalog No. 50-311-2936 Supplier Selleck Chemical LLC Supplier No. S25551G
Clarithromycin (A-56268) is a macrolide antibiotic and a CYP3A4 inhibitor used to treat a number of bacterial infections