Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clarithromycin, G-Biosciences
SCI_ED

Catalog No. S880114
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S880114 1 g
S000027 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S880114 Supplier G-Biosciences Supplier No. RC975
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clarithromycin is a macrolide antibiotic and is an acid-stable form of erythromycin. It is effective against gram-negative and gram-positive bacteria.

Clarithromycin inhibits bacterial protein synthesis by binding to the 50S ribosomal subunit, preventing bacteria from growing and multiplying.

Chemical Identifiers

CAS 81103-11-9

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 1 g
Percent Purity 98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.