Learn More
Sigma Aldrich Fine Chemicals Biosciences Clarithromycin, Ready Made Solution 50 mg/mL in DMSO, 0.2 mum filtered | 81103-11-9 | MFCD00865140 | 2ML
Catalog No. 501758302
Encompass
Clarithromycin, Ready Made Solution 50 mg/mL in DMSO, 0.2 mum filtered | Mol Wt: 747.95 | 81103-11-9 | MFCD00865140 | 2ML
Catalog No. 50-175-8302 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SBR000112ML
Clarithromycin, Ready Made Solution 50 mg/mL in DMSO, 0.2 mum filtered | Mol Wt: 747.95 | 81103-11-9 | MFCD00865140 | 2ML