Learn More
Medchemexpress LLC Clarithromycin (Standard) | 81103-11-9 | 99.9% | 747.95 | 100 MG
SDP
Catalog No. 5000386385
Encompass_Preferred
Clarithromycin (Standard) is the analytical standard of Clarithromycin. This product is intended for research and analytical applications. It has a broad spectrum of antimicrobial activity and inhibits the CYP3A4-catalyzed triazolam alpha-hydroxylation. Clarithromycin also significantly inhibits the HERG potassium current and affects the autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • The compound is an analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: Solid.
  • Color: White to off-white.
Catalog No. 50-003-86385 Supplier Medchemexpress LLC Supplier No. HY17508R100MG
Clarithromycin (Standard) is the analytical standard of Clarithromycin. This product is intended for research and analytical applications. It has a broad spectrum of antimicrobial activity and inhibits the CYP3A4-catalyzed triazolam alpha-hydroxylation. Clarithromycin also significantly inhibits the HERG potassium current and affects the autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • The compound is an analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: Solid.
  • Color: White to off-white.