Learn More
Medchemexpress LLC Clarithromycin (Standard) | 81103-11-9 | 99.9% | 747.95 | 25 MG
SDP
Catalog No. 5000384454
Encompass_Preferred
Clarithromycin (Standard) is the analytical standard of Clarithromycin, intended for research and analytical applications. It exhibits a broad spectrum of antimicrobial activity. It inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation with an IC50 (Ki) value of 56 (43) μM and significantly inhibits the HERG potassium current. Clarithromycin affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • Intended for research and analytical applications.
  • Exhibits a broad spectrum of antimicrobial activity.
  • Inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation.
  • Significantly inhibits the HERG potassium current.
  • Affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-84454 Supplier Medchemexpress LLC Supplier No. 25MGHY17508R
Clarithromycin (Standard) is the analytical standard of Clarithromycin, intended for research and analytical applications. It exhibits a broad spectrum of antimicrobial activity. It inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation with an IC50 (Ki) value of 56 (43) μM and significantly inhibits the HERG potassium current. Clarithromycin affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • Intended for research and analytical applications.
  • Exhibits a broad spectrum of antimicrobial activity.
  • Inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation.
  • Significantly inhibits the HERG potassium current.
  • Affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.