Learn More
Medchemexpress LLC Clarithromycin (Standard) | 81103-11-9 | 99.9% | 747.95 | 50 MG
SDP
Catalog No. 5000386311
Encompass_Preferred
Clarithromycin (Standard) is the analytical standard of Clarithromycin, intended for research and analytical applications. It possesses a broad spectrum of antimicrobial activity and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation with an IC50 (Ki) value of 56 (43) μM.
  • Significantly inhibits the hERG potassium current.
  • Affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
Catalog No. 50-003-86311 Supplier Medchemexpress LLC Supplier No. HY17508R50MG
Clarithromycin (Standard) is the analytical standard of Clarithromycin, intended for research and analytical applications. It possesses a broad spectrum of antimicrobial activity and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation with an IC50 (Ki) value of 56 (43) μM.
  • Significantly inhibits the hERG potassium current.
  • Affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.