Learn More
Electron Microscopy Sciences Class 10 Sticky Swabs 1 mm head
SDP
Catalog No. 501930628
Encompass_Preferred
Dust-free sticky swabs are specially used and designed for a high degree dust-free environment (class10-100). Anti-static treatment can safely dissipate electrostatic charge. Sticky swabs are applicable for manufacturing and assembling static sensitive products.
Catalog No. 50-193-0628 Supplier Electron Microscopy Sciences Supplier No. 7832573
Dust-free sticky swabs are specially used and designed for a high degree dust-free environment (class10-100). Anti-static treatment can safely dissipate electrostatic charge. Sticky swabs are applicable for manufacturing and assembling static sensitive products.