Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Class 3.1 Independent Temperature Safety Device
SDP

Catalog No. 08111630 Shop All BINDER Inc. Products
Catalog No. 08-111-630 Supplier BINDER™ Supplier No. 80120985
Add to Cart
Add to Cart

For Use With: BINDER units ED 56, ED 115, ED 260, ED 720

  • Class 3.1
  • With visual alarm
  • Safety devices also available in Class 2 and 3.3

Specifications

Description Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
For Use With (Equipment) BINDER Chambers ED 56, ED 115, ED 260, ED 720
Material Plastic
Type Temperature Safety Device
For Use With (Application) Temperature Regulation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.