Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Class 3.1 Independent Temperature Safety Device
SDP

Catalog No. 08111632 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers FD 56/115/260/720
BINDER Chambers FED 56/115/260/720
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-111-632 BINDER Chambers FD 56/115/260/720
08-111-633 BINDER Chambers FED 56/115/260/720
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-111-632 Supplier BINDER™ Supplier No. 80121643
Add to Cart
Add to Cart

Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)

  • Class 3.1
  • With visual alarm
  • Safety devices also available in Class 2 and 3.3

Specifications

Description Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
For Use With (Equipment) BINDER Chambers FD 56/115/260/720
Material Plastic
Type Temperature Calibration Certificate
For Use With (Application) Temperature Regulation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.