Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Class 3.1
- With visual alarm
- Safety devices also available in Class 2 and 3.3
Specifications
Specifications
| Description | Class 3.1 independent temperature safety device - with visual alarm (DIN 12880) |
| For Use With (Equipment) | BINDER Chambers FED 56/115/260/720 |
| Material | Plastic |
| Type | Temperature Calibration Certificate |
| For Use With (Application) | Temperature Regulation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
