Learn More
Pyramex™ Class 3 Heat Sealed Tape T-Shirts
Catalog No. 19555440 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-440 Hi-vis Orange X-Large
19-555-428 Hi-vis Lime Medium
19-555-427 Hi-vis Lime Large
19-555-433 Hi-vis Lime X-Large
19-555-429 Hi-vis Lime 2X-Large
19-555-430 Hi-vis Lime 3X-Large
19-555-431 Hi-vis Lime 4X-Large
19-555-432 Hi-vis Lime 5X-Large
19-555-435 Hi-vis Orange Medium
19-555-434 Hi-vis Orange Large
19-555-436 Hi-vis Orange 2X-Large
19-555-437 Hi-vis Orange 3X-Large
19-555-438 Hi-vis Orange 4X-Large
19-555-439 Hi-vis Orange 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-555-440 Supplier Pyramex™ Supplier No. RTS3420XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Class 3 Heat Sealed Tape T-Shirts available in two highly visible colors
  • Hi-Vis lightweight polyester moisture wicking material
  • 2 in. heat sealed silver reflective material
  • Chest pocket measures 5.5 x 5.5 in.
Specifications
Size X-Large
Material Polyester
Color Hi-vis Orange
Sleeve Style Short Sleeve
No. of Pockets 1
Certifications/Compliance Type R - Class 3
Tape Type Heat-sealed