Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Class 3 Multi-Layer Parka

Catalog No. 19555336 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-336 Medium
19-555-335 Large
19-555-341 X-Large
19-555-337 2X-Large
19-555-338 3X-Large
19-555-339 4X-Large
19-555-340 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-555-336 Supplier Pyramex™ Supplier No. RPB3610M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pyramex™ Class 3 Multi-Layer Parka with ANSI/ISEA 107-2015 safety compliance

Outer Jacket

  • Waterproof Hi-Vis polyester shell
  • Quilted lining - Teflon™ treated
  • Elastic band at wrists
  • Concealed detachable hood with drawstring and slack adjustment
  • Interior access zipper for print customization
  • Dual zipper front closure with hook and loop storm flap
  • Badge lanyard, d-ring hook, and 2 mic tabs
  • Heat sealed seams
  • Drawstring around bottom
  • 7 outer, 5 inner pockets

Inner Jacket

  • Hi-Vis lime polyester fleece
  • 2 in. non-connected silver reflective material
  • Elastic band at wrists
  • Drawstring around bottom
  • 2 outer, 2 inner pockets

Specifications

Size Medium
Material Polyester
Color Hi-vis Lime
Closure Type Dual Zipper
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 3
Coating Teflon™ and Polyurethane
Cuff Style Elastic with scratch band
Hood Type Concealed, Detachable
Liner Material Quilted Polyester Cotton, Taffeta
No. of Pockets 16
Resistance Weatherproof
Tape Type China Brand Segmented Heat-Sealed
Size (English) Chest 38 to 40 in.
Length (English) 31.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.