Learn More
Pyramex™ Class 3 Premium Zipper Sweatshirts
Catalog No. 19555357 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-357 Hi-vis Lime Medium
19-555-356 Hi-vis Lime Large
19-555-362 Hi-vis Lime X-Large
19-555-358 Hi-vis Lime 2X-Large
19-555-359 Hi-vis Lime 3X-Large
19-555-360 Hi-vis Lime 4X-Large
19-555-361 Hi-vis Lime 5X-Large
19-555-364 Hi-vis Orange Medium
19-555-363 Hi-vis Orange Large
19-555-369 Hi-vis Orange X-Large
19-555-365 Hi-vis Orange 2X-Large
19-555-366 Hi-vis Orange 3X-Large
19-555-367 Hi-vis Orange 4X-Large
19-555-368 Hi-vis Orange 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-555-357 Supplier Pyramex™ Supplier No. RSZH3310M
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Class 3 Premium Zipper Sweatshirts with ANSI/ISEA 107-2015 safety compliance
  • Weatherproof
  • Hi-Vis Polyester material - Teflon™ treated
  • Elastic band at the waist and wrists
  • 2 in. silver reflective material
  • Chest pocket with clear panel, hook and loop closure
  • 2 mic tabs
  • Drawstring hood
  • Zipper front closure
Specifications
Size Medium
Material Teflon-treated Polyester
Color Hi-vis Lime
Sleeve Style Long Sleeve
Closure Type Zipper
Cuff Style Elastic
No. of Pockets 4
Resistance Weatherproof
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 3
Size (English) Chest 38 to 40 in.
Tape Type China Brand Silver
Length (English) 27 in.