Learn More
SPS Medical CLASS 5 STEAM STERILIZE 1000PK
Catalog No. NC0178696
Encompass
An immediate readout integrator for use in gravity, prevacuum and immediate use sterilizer cycles. Tested to ST-60 and ISO 11140-1-2005. FDA cleared as equivalent in performance to a biological indicator.
Catalog No. NC0178696 Supplier SPS Medical Supplier No. SSI1000
An immediate readout integrator for use in gravity, prevacuum and immediate use sterilizer cycles. Tested to ST-60 and ISO 11140-1-2005. FDA cleared as equivalent in performance to a biological indicator.