Learn More
Labelmaster Inc Miscellaneous Dangerous Goods Label, Paper, Roll of 500
SDP
Catalog No. NC0763606
Encompass_Preferred
Help take the guessing out of your shipping label solution with Labelmaster's Miscellaneous Dangerous Goods Labels, revised to meet the design requirements of 49 CFR 172.407. In addition to complying with directives set by the DOT, these also meet the design and durability standards of 49 CFR, ICAO and other international air regulations. You'll be worry-free as these Hazard Class 9 labels are printed with light-fast inks for fade resistance, and adhesives are formulated for use in various climates and environments as well as strong holds on corrugated boxes.
Catalog No. NC0763606 Supplier Labelmaster Inc Supplier No. HML9
Help take the guessing out of your shipping label solution with Labelmaster's Miscellaneous Dangerous Goods Labels, revised to meet the design requirements of 49 CFR 172.407. In addition to complying with directives set by the DOT, these also meet the design and durability standards of 49 CFR, ICAO and other international air regulations. You'll be worry-free as these Hazard Class 9 labels are printed with light-fast inks for fade resistance, and adhesives are formulated for use in various climates and environments as well as strong holds on corrugated boxes.