Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Class II HDAC Antibody Sampler Kit, 1 Kit
SDP

Catalog No. 502731151
Encompass

The Class II HDAC Antibody Sampler Kit provides an economical means of detecting Class II HDAC proteins using control antibodies against HDAC4, HDAC5, HDAC6, and HDAC7. The kit contains enough primary antibodies to perform at least two western blot experiments.

  • Contains primary antibodies against HDAC4, HDAC5, HDAC6, and HDAC7
  • Includes secondary anti-rabbit IgG, HRP-linked antibody
  • Designed for western blotting applications
  • Sufficient for at least two western blot experiments

Catalog No. 50-273-1151 Supplier Cell Signaling Technology Supplier No. 79891T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Class II HDAC Antibody Sampler Kit provides an economical means of detecting Class II HDAC proteins using control antibodies against HDAC4, HDAC5, HDAC6, and HDAC7. The kit contains enough primary antibodies to perform at least two western blot experiments.

  • Contains primary antibodies against HDAC4, HDAC5, HDAC6, and HDAC7
  • Includes secondary anti-rabbit IgG, HRP-linked antibody
  • Designed for western blotting applications
  • Sufficient for at least two western blot experiments

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.