Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0625 in. Diameter, 2 in. Length, 4 Microinch Finish, 1 ea.
Catalog No. 5000516134
Encompass
This Class X standard plug gage is designed for precise measurement applications, including the calibration of measuring equipment or gaging parts directly on the shop floor and in metrology laboratories. Constructed from tool steel with a hardness of Rc 60/65, the gage features a durable chrome-plated finish for enhanced longevity and accuracy.
  • Diameter of 0.0625 in.
  • Overall length of 2 in.
  • Tolerance of -0.00004 in.
  • Surface finish of 4 microinch or better.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ISO 17025 standards.
Catalog No. 50-005-16134 Supplier MSC Supplier No. 88806252
This Class X standard plug gage is designed for precise measurement applications, including the calibration of measuring equipment or gaging parts directly on the shop floor and in metrology laboratories. Constructed from tool steel with a hardness of Rc 60/65, the gage features a durable chrome-plated finish for enhanced longevity and accuracy.
  • Diameter of 0.0625 in.
  • Overall length of 2 in.
  • Tolerance of -0.00004 in.
  • Surface finish of 4 microinch or better.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ISO 17025 standards.