Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.4610 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000515878
Encompass
This Class X Plus pin gage is designed for precision calibration of measuring equipment or direct gaging of parts. It is constructed from tool steel and features a cold-stabilized finish for durability and accuracy.
  • Diameter of 0.4610 in.
  • Length of 2 in.
  • Class X Plus tolerance of 0.00004 in.
  • Tolerance range of -0.00000 / +0.00004 in.
  • Hardness rating of Rc 60-62
  • Roundness within 0.00002 in.
  • Bright, uncoated finish of 2 microinch
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-15878 Supplier MSC Supplier No. 88746102
This Class X Plus pin gage is designed for precision calibration of measuring equipment or direct gaging of parts. It is constructed from tool steel and features a cold-stabilized finish for durability and accuracy.
  • Diameter of 0.4610 in.
  • Length of 2 in.
  • Class X Plus tolerance of 0.00004 in.
  • Tolerance range of -0.00000 / +0.00004 in.
  • Hardness rating of Rc 60-62
  • Roundness within 0.00002 in.
  • Bright, uncoated finish of 2 microinch
  • Includes a certificate of accuracy with NIST traceability