Learn More
MSC Vermont Gage Plug Gage, Steel, 0.1560 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000547961
Encompass
This Class X Plus pin gage is designed for precision measurements, including the calibration of measuring equipment and direct gaging of parts. It features a 0.1560 in. diameter and a 2 in. length, constructed from durable tool steel.
  • Tolerance of 0.00004 in.
  • Hardness of 60 to 65 Rockwell
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Uncoated finish
Catalog No. 50-005-47961 Supplier MSC Supplier No. 88715602
This Class X Plus pin gage is designed for precision measurements, including the calibration of measuring equipment and direct gaging of parts. It features a 0.1560 in. diameter and a 2 in. length, constructed from durable tool steel.
  • Tolerance of 0.00004 in.
  • Hardness of 60 to 65 Rockwell
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Uncoated finish