Learn More
MSC Vermont Gage Class X Plus Pin Gage, Tool Steel, 0.0590 in. Diameter, 2 in. Long, 1 ea.
Catalog No. 5000547869
Encompass
This Class X Plus pin gage is a high-precision measurement tool designed for the calibration of measuring equipment or for gauging parts directly. Constructed from durable tool steel, this pin gage offers consistent reliability and wear resistance for industrial inspection requirements.
  • Manufactured from tool steel
  • Diameter of 0.0590 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Surface finish of 4 micro
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-47869 Supplier MSC Supplier No. 88705900
This Class X Plus pin gage is a high-precision measurement tool designed for the calibration of measuring equipment or for gauging parts directly. Constructed from durable tool steel, this pin gage offers consistent reliability and wear resistance for industrial inspection requirements.
  • Manufactured from tool steel
  • Diameter of 0.0590 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Surface finish of 4 micro
  • Includes certificate of accuracy with NIST traceability