Learn More
MSC Vermont Gage Plug Gage, Class X Plus, 0.0700 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000511813
Encompass
This Class X Plus pin gage is a precision-machined tool designed for the calibration of measuring equipment or for gauging parts directly. Constructed from tool steel with a hardness of Rockwell 60/65, it provides durability and reliability for demanding inspection environments.
  • Diameter of 0.0700 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Surface finish of 4 microinches or better
  • Includes a certificate of accuracy with NIST traceability
  • Round within 0.00002 in.
Catalog No. 50-005-11813 Supplier MSC Supplier No. 88707005
This Class X Plus pin gage is a precision-machined tool designed for the calibration of measuring equipment or for gauging parts directly. Constructed from tool steel with a hardness of Rockwell 60/65, it provides durability and reliability for demanding inspection environments.
  • Diameter of 0.0700 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Surface finish of 4 microinches or better
  • Includes a certificate of accuracy with NIST traceability
  • Round within 0.00002 in.