Learn More
MSC Vermont Gage Class X Pin Gauge, 0.2980 in., Steel, 1 ea.
Catalog No. 5000511825
Encompass
Standard Class X plug gages serve as precise masters for calibrating measuring equipment or as tools for direct part inspection. These gages are engineered to provide accurate, cost-effective measurement standards suitable for both shop floor environments and metrology laboratories.
  • Constructed from tool steel hardened to 60/65 Rc
  • Length of 2 in.
  • Surface finish of 4 microinches or better
  • Roundness within 1/2 of the diameter tolerance
  • Includes a certificate of accuracy with NIST traceability
  • Tolerances maintained at +0.00004 in. for sizes up to 0.8250 in.
Catalog No. 50-005-11825 Supplier MSC Supplier No. 88729801
Standard Class X plug gages serve as precise masters for calibrating measuring equipment or as tools for direct part inspection. These gages are engineered to provide accurate, cost-effective measurement standards suitable for both shop floor environments and metrology laboratories.
  • Constructed from tool steel hardened to 60/65 Rc
  • Length of 2 in.
  • Surface finish of 4 microinches or better
  • Roundness within 1/2 of the diameter tolerance
  • Includes a certificate of accuracy with NIST traceability
  • Tolerances maintained at +0.00004 in. for sizes up to 0.8250 in.