Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0110 in. Diameter, 2 in. Length, 4 Micro Finish, 1 ea.
Catalog No. 5000511765
Encompass
This Class X minus pin gage is a precision tool used for the calibration of measuring equipment or for gaging parts directly. Constructed from durable tool steel, it features a diameter of 0.0110 in. and a total length of 2 in.
  • Class X tolerance of -0.00004 in.
  • Hardened tool steel construction
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Made in the United States
Catalog No. 50-005-11765 Supplier MSC Supplier No. 88801105
This Class X minus pin gage is a precision tool used for the calibration of measuring equipment or for gaging parts directly. Constructed from durable tool steel, it features a diameter of 0.0110 in. and a total length of 2 in.
  • Class X tolerance of -0.00004 in.
  • Hardened tool steel construction
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Made in the United States