Learn More
MSC Vermont Gage Plug Gage, Class X, Tool Steel, 0.0920 in. Diameter, 2 in. Length, 1/Pk.
Catalog No. 5000511775
Encompass
This Class X plug gage is designed for high-precision calibration of measuring equipment or for direct inspection of parts. Manufactured from tool steel with a Rockwell hardness of 60/65, it provides excellent durability and consistent performance in industrial metrology applications. This gage includes a certificate of accuracy with NIST traceability to ensure compliance with quality standards.
  • Class X tolerance of 0.00004 in.
  • Hardened tool steel construction
  • Surface finish of four microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
  • Nominal diameter of 0.0920 in.
  • Overall length of 2 in.
Catalog No. 50-005-11775 Supplier MSC Supplier No. 88809207
This Class X plug gage is designed for high-precision calibration of measuring equipment or for direct inspection of parts. Manufactured from tool steel with a Rockwell hardness of 60/65, it provides excellent durability and consistent performance in industrial metrology applications. This gage includes a certificate of accuracy with NIST traceability to ensure compliance with quality standards.
  • Class X tolerance of 0.00004 in.
  • Hardened tool steel construction
  • Surface finish of four microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
  • Nominal diameter of 0.0920 in.
  • Overall length of 2 in.