Learn More
MSC Vermont Gage Go Plug Gage, Class X, Tool Steel, 0.7520 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000511306
Encompass
This Class X standard plug gage is a high-precision tool designed for the calibration of measuring equipment or for direct inspection of parts. Engineered for accuracy and durability, it features a 0.7520 in. diameter and a 2 in. overall length.
  • Hardened tool steel construction with a surface finish of 4 microinch or better for enhanced wear resistance.
  • Tolerance of 0.00004 in. ensuring precise measurement capabilities.
  • Includes a certificate of accuracy with NIST traceability to meet ISO 17025 standards.
  • Reversible style design extends the useful life of the gage.
  • Roundness within 1/2 of tolerance.
Catalog No. 50-005-11306 Supplier MSC Supplier No. 88775200
This Class X standard plug gage is a high-precision tool designed for the calibration of measuring equipment or for direct inspection of parts. Engineered for accuracy and durability, it features a 0.7520 in. diameter and a 2 in. overall length.
  • Hardened tool steel construction with a surface finish of 4 microinch or better for enhanced wear resistance.
  • Tolerance of 0.00004 in. ensuring precise measurement capabilities.
  • Includes a certificate of accuracy with NIST traceability to meet ISO 17025 standards.
  • Reversible style design extends the useful life of the gage.
  • Roundness within 1/2 of tolerance.