Learn More
MSC Vermont Gage Class X Minus Plug Gage, Tool Steel, 0.1610 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000511316
Encompass
This Class X standard plug gage is designed for precise calibration of measuring equipment or for direct gaging of parts. It features a minus tolerance and is engineered to provide reliable inspection performance in industrial measurement applications.
  • Diameter of 0.1610 in.
  • Overall length of 2 in.
  • Constructed from tool steel with a hardness of Rc 60 to 65
  • Surface finish of 4 microinch or better
  • Tolerance of 0.00004 in.
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
Catalog No. 50-005-11316 Supplier MSC Supplier No. 88816103
This Class X standard plug gage is designed for precise calibration of measuring equipment or for direct gaging of parts. It features a minus tolerance and is engineered to provide reliable inspection performance in industrial measurement applications.
  • Diameter of 0.1610 in.
  • Overall length of 2 in.
  • Constructed from tool steel with a hardness of Rc 60 to 65
  • Surface finish of 4 microinch or better
  • Tolerance of 0.00004 in.
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards