Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.4725 in. Diameter, 2 in. Long, 1 ea.
Catalog No. 5000511318
Encompass
This Class X minus pin gage is designed for the calibration of measuring equipment or for gaging parts directly. Constructed from tool steel, this gage provides high-precision measurement capabilities for metalworking and inspection applications.
  • Diameter of 0.4725 in.
  • Overall length of 2 in.
  • Tolerance of +0.0000 in., -0.00004 in.
  • Hardness rating of 60/65 Rc
  • Surface finish of 4 microinch or better
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-11318 Supplier MSC Supplier No. 88847256
This Class X minus pin gage is designed for the calibration of measuring equipment or for gaging parts directly. Constructed from tool steel, this gage provides high-precision measurement capabilities for metalworking and inspection applications.
  • Diameter of 0.4725 in.
  • Overall length of 2 in.
  • Tolerance of +0.0000 in., -0.00004 in.
  • Hardness rating of 60/65 Rc
  • Surface finish of 4 microinch or better
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability