Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.9680 in. Diameter, 2 in. Long, Steel, 1 ea.
Catalog No. 5000511260
Encompass
This Class X Plus pin gage is designed for the calibration of measuring equipment or for direct gaging of parts. Constructed from steel with a bright, uncoated finish, this precision tool provides a diameter of 0.9680 in. and a length of 2 in.
  • Tolerance of 0.00006 in.
  • Round within 0.5 of tolerance
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-11260 Supplier MSC Supplier No. 88796800
This Class X Plus pin gage is designed for the calibration of measuring equipment or for direct gaging of parts. Constructed from steel with a bright, uncoated finish, this precision tool provides a diameter of 0.9680 in. and a length of 2 in.
  • Tolerance of 0.00006 in.
  • Round within 0.5 of tolerance
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability