Learn More
MSC Vermont Gage Plug Gage, Class X, Tool Steel, 0.1615 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000511201
Encompass
These Class X plug gages are designed for precision measurements on the shop floor or in the metrology laboratory. They serve as reliable masters for calibrating measuring equipment or for direct gaging of parts. Each gage is constructed from durable tool steel and features a high-quality surface finish to ensure consistent performance.
  • Accuracy of 0.00004 in.
  • Hardness rated at Rockwell 60 to 65
  • Diameter of 0.1615 in. with an overall length of 2 in.
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Round within half of the tolerance
Catalog No. 50-005-11201 Supplier MSC Supplier No. 88816152
These Class X plug gages are designed for precision measurements on the shop floor or in the metrology laboratory. They serve as reliable masters for calibrating measuring equipment or for direct gaging of parts. Each gage is constructed from durable tool steel and features a high-quality surface finish to ensure consistent performance.
  • Accuracy of 0.00004 in.
  • Hardness rated at Rockwell 60 to 65
  • Diameter of 0.1615 in. with an overall length of 2 in.
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Round within half of the tolerance