Learn More
MSC Vermont Gage Pin Gage, Class X, Minus, 0.1410 in. Diameter, 2 in. Long, 1 ea.
Catalog No. 5000511203
Encompass
This Class X minus pin gage is a precision measurement tool with a diameter of 0.1410 in. and a length of 2 in., designed for the calibration of measuring equipment or for direct parts inspection. Constructed from tool steel and treated for durability, it provides high-accuracy measurement capabilities for industrial metrology applications.
  • Tolerance of -0.00004 in.
  • Surface finish of 4 microinch or better
  • Hardness rated at 60/65 RC
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Reversible style for extended service life
Catalog No. 50-005-11203 Supplier MSC Supplier No. 88814108
This Class X minus pin gage is a precision measurement tool with a diameter of 0.1410 in. and a length of 2 in., designed for the calibration of measuring equipment or for direct parts inspection. Constructed from tool steel and treated for durability, it provides high-accuracy measurement capabilities for industrial metrology applications.
  • Tolerance of -0.00004 in.
  • Surface finish of 4 microinch or better
  • Hardness rated at 60/65 RC
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Reversible style for extended service life