Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.8755 in. Diameter, 2 in. Long, Tool Steel, 1 ea.
Catalog No. 5000511206
Encompass
This Class X No-Go (Minus) plug gage is designed for high-precision measurement tasks, including the calibration of measuring equipment and direct part gaging. Constructed from durable tool steel, it features a 0.8755 in. diameter and a 2 in. length, manufactured to meet rigorous inspection standards.
  • Tool steel construction
  • Hardness rated at RC 60/65
  • Surface finish of 4 microinch or better
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-11206 Supplier MSC Supplier No. 88887559
This Class X No-Go (Minus) plug gage is designed for high-precision measurement tasks, including the calibration of measuring equipment and direct part gaging. Constructed from durable tool steel, it features a 0.8755 in. diameter and a 2 in. length, manufactured to meet rigorous inspection standards.
  • Tool steel construction
  • Hardness rated at RC 60/65
  • Surface finish of 4 microinch or better
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability