Learn More
MSC Vermont Gage Standard Plug Gage, Class X, Tool Steel, 0.0305 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000510988
Encompass
This Class X standard plug gage is designed for precision inspection tasks in both shop floor and metrology laboratory environments. It is utilized as a master for calibrating measuring equipment or for gaging parts directly to ensure dimensional accuracy. Constructed from durable tool steel, the gage is built for consistent performance in demanding industrial applications.
  • Diameter of 0.0305 in. with a tolerance of +0.00004 in.
  • Hardness rating of 60/65 Rc for increased wear resistance
  • Overall length of 2 in.
  • Surface finish of 4 microinch or better
  • Round within 0.5 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
Catalog No. 50-005-10988 Supplier MSC Supplier No. 88703053
This Class X standard plug gage is designed for precision inspection tasks in both shop floor and metrology laboratory environments. It is utilized as a master for calibrating measuring equipment or for gaging parts directly to ensure dimensional accuracy. Constructed from durable tool steel, the gage is built for consistent performance in demanding industrial applications.
  • Diameter of 0.0305 in. with a tolerance of +0.00004 in.
  • Hardness rating of 60/65 Rc for increased wear resistance
  • Overall length of 2 in.
  • Surface finish of 4 microinch or better
  • Round within 0.5 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards