Learn More
MSC Vermont Gage Class X No-Go Pin Gage, Tool Steel, 0.1000 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000510466
Encompass
This Class X No-Go (Minus) pin gage is a precision tool designed for the calibration of measuring equipment or for direct gaging of parts. Featuring a 0.1000 in. diameter and a 2 in. length, this gage is constructed from tool steel to ensure durability and reliability in inspection applications.
  • Tolerance of -0.00004 in.
  • Hardness rating of 60/65 HRC
  • Finish of 4 microinch or better
  • Round within 1/2 of tolerance
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-10466 Supplier MSC Supplier No. 88810007
This Class X No-Go (Minus) pin gage is a precision tool designed for the calibration of measuring equipment or for direct gaging of parts. Featuring a 0.1000 in. diameter and a 2 in. length, this gage is constructed from tool steel to ensure durability and reliability in inspection applications.
  • Tolerance of -0.00004 in.
  • Hardness rating of 60/65 HRC
  • Finish of 4 microinch or better
  • Round within 1/2 of tolerance
  • Includes a certificate of accuracy with NIST traceability