Learn More
MSC Vermont Gage Plug Gage, Tool Steel, 0.8755 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000510430
Encompass
This Class X Plus plug gage is designed for precision measurements, serving as an essential tool for calibrating measuring equipment or gauging parts directly. Constructed from tool steel and cold stabilized to a hardness of 60 to 65 Rc, this gage ensures durability and consistent performance in demanding environments.
  • Features a nominal diameter of 0.8755 in. and an overall length of 2 in.
  • Includes a certificate of accuracy with NIST traceability to ensure reliable measurement standards.
  • Maintains a surface finish of 4 microinch or better for precise contact.
  • Engineered with a tolerance of 0.00004 in. for high-accuracy applications.
  • Designed to be round within one-half of the tolerance.
Catalog No. 50-005-10430 Supplier MSC Supplier No. 88787551
This Class X Plus plug gage is designed for precision measurements, serving as an essential tool for calibrating measuring equipment or gauging parts directly. Constructed from tool steel and cold stabilized to a hardness of 60 to 65 Rc, this gage ensures durability and consistent performance in demanding environments.
  • Features a nominal diameter of 0.8755 in. and an overall length of 2 in.
  • Includes a certificate of accuracy with NIST traceability to ensure reliable measurement standards.
  • Maintains a surface finish of 4 microinch or better for precise contact.
  • Engineered with a tolerance of 0.00004 in. for high-accuracy applications.
  • Designed to be round within one-half of the tolerance.