Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Vermont Gage Pin Gage, Class X, Tool Steel, 0.0850 in. Diameter, 2 in. Length, 1 ea

Catalog No. 5000510319
Encompass

This Class X Plus plug gage is a high-precision tool steel reference standard designed for accurate inspection and calibration tasks. Engineered for stability and reliability, it serves as a master gage for calibrating measuring equipment such as micrometers and snap gages, or for direct pass/fail inspection of manufactured parts.

  • Diameter: 0.0850 in.
  • Overall length: 2 in.
  • Tolerance: +0.00004 in. / -0.00000 in.
  • Hardness: RC 60 to 65
  • Material: Cold stabilized tool steel
  • Surface finish: 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Suitable for high-volume inspection and calibration applications

Catalog No. 50-005-10319 Supplier MSC Supplier No. 88708508
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Class X Plus plug gage is a high-precision tool steel reference standard designed for accurate inspection and calibration tasks. Engineered for stability and reliability, it serves as a master gage for calibrating measuring equipment such as micrometers and snap gages, or for direct pass/fail inspection of manufactured parts.

  • Diameter: 0.0850 in.
  • Overall length: 2 in.
  • Tolerance: +0.00004 in. / -0.00000 in.
  • Hardness: RC 60 to 65
  • Material: Cold stabilized tool steel
  • Surface finish: 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Suitable for high-volume inspection and calibration applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.