Learn More
MSC Vermont Gage Class X Plus Pin Gage, Tool Steel, 0.2060 in. Diameter, 2 in. Long, 1 ea.
Catalog No. 5000510326
Encompass
This Class X Plus pin gage is designed for precision measurements, serving as an essential tool for calibrating measuring equipment or checking parts directly. Manufactured from cold-stabilized tool steel hardened to Rc 60-62, this gage provides high durability and consistent performance in demanding industrial inspection environments.
  • Diameter of 0.2060 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Single tolerance of -.00000 in. / +.00004 in.
  • Round within 0.00002 in.
  • Surface finish of 4 micro
  • Includes certificate of accuracy with NIST traceability
  • Constructed from cold-stabilized tool steel
  • Made in the United States of America
Catalog No. 50-005-10326 Supplier MSC Supplier No. 88720602
This Class X Plus pin gage is designed for precision measurements, serving as an essential tool for calibrating measuring equipment or checking parts directly. Manufactured from cold-stabilized tool steel hardened to Rc 60-62, this gage provides high durability and consistent performance in demanding industrial inspection environments.
  • Diameter of 0.2060 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Single tolerance of -.00000 in. / +.00004 in.
  • Round within 0.00002 in.
  • Surface finish of 4 micro
  • Includes certificate of accuracy with NIST traceability
  • Constructed from cold-stabilized tool steel
  • Made in the United States of America