Learn More
MSC Vermont Gage Minus Pin Gage, Class X, 0.7500 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000510096
Encompass
This Class X minus pin gage is designed for the precise calibration of measuring equipment or for direct inspection of parts. Manufactured from tool steel, it provides a durable, uncoated finish for reliable use in dimensional metrology.
  • Class X tolerance of 0.00004 in.
  • Diameter of 0.7500 in.
  • Overall length of 2 in.
  • Hardness rating of 60/65 Rc
  • Surface finish of 4 microinch or better
  • Round within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
Catalog No. 50-005-10096 Supplier MSC Supplier No. 88875000
This Class X minus pin gage is designed for the precise calibration of measuring equipment or for direct inspection of parts. Manufactured from tool steel, it provides a durable, uncoated finish for reliable use in dimensional metrology.
  • Class X tolerance of 0.00004 in.
  • Diameter of 0.7500 in.
  • Overall length of 2 in.
  • Hardness rating of 60/65 Rc
  • Surface finish of 4 microinch or better
  • Round within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards