Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.025 in. Diameter, Tool Steel, 2 in. Length, 1 ea.
Catalog No. 5000510105
Encompass
This Class X No-Go (Minus) pin gage is designed for the direct gaging of parts or as a master for calibrating measuring equipment. Featuring a 0.025 in. diameter, this gage is constructed from tool steel and hardened to Rc 60/65 for durability in both shop floor and metrology laboratory environments.
  • Manufactured from tool steel with a hardness of Rc 60/65
  • Provides a 4 microinch finish or better
  • Includes a certificate of accuracy with NIST traceability
  • Maintains roundness within 1/2 of the tolerance
  • Meets ISO 17025 standards
  • Class X tolerance of 0.00004 in.
  • Overall length of 2 in.
Catalog No. 50-005-10105 Supplier MSC Supplier No. 88802509
This Class X No-Go (Minus) pin gage is designed for the direct gaging of parts or as a master for calibrating measuring equipment. Featuring a 0.025 in. diameter, this gage is constructed from tool steel and hardened to Rc 60/65 for durability in both shop floor and metrology laboratory environments.
  • Manufactured from tool steel with a hardness of Rc 60/65
  • Provides a 4 microinch finish or better
  • Includes a certificate of accuracy with NIST traceability
  • Maintains roundness within 1/2 of the tolerance
  • Meets ISO 17025 standards
  • Class X tolerance of 0.00004 in.
  • Overall length of 2 in.