Learn More
MSC Vermont Gage Class X Plus Pin Gage, Tool Steel, 0.2520 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000516407
Encompass
This Class X Plus pin gage is designed for precise calibration of measuring equipment and direct inspection of parts. Manufactured from tool steel, this pin gage offers high durability and reliability for demanding metrology applications.
  • Diameter of 0.2520 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Hardness rating of 60 to 65 Rc, cold stabilized
  • Surface finish of 4 microinch or better
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
Catalog No. 50-005-16407 Supplier MSC Supplier No. 88725205
This Class X Plus pin gage is designed for precise calibration of measuring equipment and direct inspection of parts. Manufactured from tool steel, this pin gage offers high durability and reliability for demanding metrology applications.
  • Diameter of 0.2520 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Hardness rating of 60 to 65 Rc, cold stabilized
  • Surface finish of 4 microinch or better
  • Roundness within 1/2 of tolerance
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel