Learn More
MSC Vermont Gage Class X Minus Plug Gage, 0.0260 in., 2 in. Long, 1 ea.
Catalog No. 5000516379
Encompass
This Class X minus pin gage is a precision-machined tool designed for the calibration of measuring equipment or for direct gaging of parts. It features a 0.0260 in. diameter and a 2 in. overall length, providing reliable measurement accuracy for critical inspection tasks.
  • Manufactured from tool steel with a hardness of 60 to 65 Rc
  • Cold stabilized for enhanced dimensional stability
  • Surface finish of 4 microinch or better
  • Round within 1/2 of tolerance
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-16379 Supplier MSC Supplier No. 88802608
This Class X minus pin gage is a precision-machined tool designed for the calibration of measuring equipment or for direct gaging of parts. It features a 0.0260 in. diameter and a 2 in. overall length, providing reliable measurement accuracy for critical inspection tasks.
  • Manufactured from tool steel with a hardness of 60 to 65 Rc
  • Cold stabilized for enhanced dimensional stability
  • Surface finish of 4 microinch or better
  • Round within 1/2 of tolerance
  • Includes a certificate of accuracy with NIST traceability