Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.473 in., 2 in. Length, 4 Micro Finish, 1 ea
Catalog No. 5000516351
Encompass
This Class X minus pin gage is designed for precision measurements, serving as an essential tool for the calibration of measuring equipment or for gauging parts directly. Constructed from durable tool steel, this gage provides reliable performance in industrial inspection environments.
  • Diameter of 0.473 in.
  • Overall length of 2 in.
  • Tolerance of -0.00004 in.
  • Surface finish of 4 micro
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-16351 Supplier MSC Supplier No. 88847306
This Class X minus pin gage is designed for precision measurements, serving as an essential tool for the calibration of measuring equipment or for gauging parts directly. Constructed from durable tool steel, this gage provides reliable performance in industrial inspection environments.
  • Diameter of 0.473 in.
  • Overall length of 2 in.
  • Tolerance of -0.00004 in.
  • Surface finish of 4 micro
  • Includes a certificate of accuracy with NIST traceability