Learn More
MSC Vermont Gage Plug Gage, Class X, Tool Steel, 0.4760 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000516195
Encompass
This Class X plug gage is designed for high-precision calibration of measuring equipment or for direct inspection of parts. Manufactured from cold-stabilized tool steel hardened to 60/65 Rc, this pin gage offers exceptional durability and wear resistance in demanding industrial environments.
  • Diameter of 0.4760 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Roundness within 1/2 of tolerance
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-16195 Supplier MSC Supplier No. 88847603
This Class X plug gage is designed for high-precision calibration of measuring equipment or for direct inspection of parts. Manufactured from cold-stabilized tool steel hardened to 60/65 Rc, this pin gage offers exceptional durability and wear resistance in demanding industrial environments.
  • Diameter of 0.4760 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Roundness within 1/2 of tolerance
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability