Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0700 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000516143
Encompass
This Class X minus pin gage is designed for precise calibration of measuring equipment or for gaging parts directly. Constructed from durable tool steel, this pin gage offers reliable performance for inspection and quality control applications.
  • Diameter of 0.0700 in.
  • Tolerance of -0.00004 in.
  • Length of 2 in.
  • Surface finish of 4 micro
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-16143 Supplier MSC Supplier No. 88807003
This Class X minus pin gage is designed for precise calibration of measuring equipment or for gaging parts directly. Constructed from durable tool steel, this pin gage offers reliable performance for inspection and quality control applications.
  • Diameter of 0.0700 in.
  • Tolerance of -0.00004 in.
  • Length of 2 in.
  • Surface finish of 4 micro
  • Includes a certificate of accuracy with NIST traceability