Learn More
MSC Vermont Gage Plug Gage, Class X, 0.2080 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000516146
Encompass
This Class X plug gage provides precision measurement for the calibration of equipment or direct gauging of parts. It features an uncoated tool steel construction with a nominal diameter of 0.2080 in. and an overall length of 2 in.
  • Accuracy of 0.00004 in.
  • Tolerance of -0.00004 in.
  • Surface finish of four microinch or better
  • Hardness rating of Rockwell 60 to 65
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-16146 Supplier MSC Supplier No. 88820808
This Class X plug gage provides precision measurement for the calibration of equipment or direct gauging of parts. It features an uncoated tool steel construction with a nominal diameter of 0.2080 in. and an overall length of 2 in.
  • Accuracy of 0.00004 in.
  • Tolerance of -0.00004 in.
  • Surface finish of four microinch or better
  • Hardness rating of Rockwell 60 to 65
  • Includes certificate of accuracy with NIST traceability