Learn More
MSC Vermont Gage Class X Minus Pin Gage, Tool Steel, 0.6245 in. Dia., 2 in. Length, 1 ea.
Catalog No. 5000516154
Encompass
This Class X minus pin gage is a precision measurement tool designed for the calibration of equipment or direct part inspection. Manufactured from cold-stabilized tool steel, this gage offers high durability and reliability for metrology applications.
  • Diameter of 0.6245 in. with a tolerance of -0.00004 in.
  • Length of 2 in.
  • Constructed from tool steel with 60/65 Rc hardness.
  • Surface finish of 4 microinch or better.
  • Roundness within 1/2 of tolerance.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-16154 Supplier MSC Supplier No. 88862453
This Class X minus pin gage is a precision measurement tool designed for the calibration of equipment or direct part inspection. Manufactured from cold-stabilized tool steel, this gage offers high durability and reliability for metrology applications.
  • Diameter of 0.6245 in. with a tolerance of -0.00004 in.
  • Length of 2 in.
  • Constructed from tool steel with 60/65 Rc hardness.
  • Surface finish of 4 microinch or better.
  • Roundness within 1/2 of tolerance.
  • Includes certificate of accuracy with NIST traceability.