Learn More
MSC Vermont Gage Class X Pin Gage, 0.2510 in. Diameter, Tool Steel, 2 in. Length, 1 ea.
Catalog No. 5000516155
Encompass
This Class X Plus plug gage is designed for precision measurements, suitable for both shop floor use and metrology laboratory applications. It serves as a reliable master for the calibration of measuring equipment or for direct part gaging, featuring an included certificate of accuracy with NIST traceability to ensure quality and reliability.
  • Tool steel construction with a hardness of 60/65 Rc
  • Cold stabilized for dimensional consistency
  • Surface finish of 4 microinch or better
  • Tolerance of +0.00004 in., -0.0000 in.
  • Overall length of 2 in.
  • Round within 1/2 of tolerance
  • Class X precision accuracy
Catalog No. 50-005-16155 Supplier MSC Supplier No. 88725106
This Class X Plus plug gage is designed for precision measurements, suitable for both shop floor use and metrology laboratory applications. It serves as a reliable master for the calibration of measuring equipment or for direct part gaging, featuring an included certificate of accuracy with NIST traceability to ensure quality and reliability.
  • Tool steel construction with a hardness of 60/65 Rc
  • Cold stabilized for dimensional consistency
  • Surface finish of 4 microinch or better
  • Tolerance of +0.00004 in., -0.0000 in.
  • Overall length of 2 in.
  • Round within 1/2 of tolerance
  • Class X precision accuracy