Learn More
MSC Vermont Gage Class X Minus Pin Gage, Steel, 0.1257 in., 2 in. Long, 1 ea.
Catalog No. 5000516951
Encompass
This Class X plug gage is a high-precision tool designed for the calibration of measuring equipment or for gauging parts directly. Constructed from cold-stabilized tool steel, the gage offers durability and consistency for demanding inspection applications.
  • Diameter of 0.1257 in.
  • Overall length of 2 in.
  • Tolerance class X
  • Hardness of 60/65 Rc
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
Catalog No. 50-005-16951 Supplier MSC Supplier No. 08541187
This Class X plug gage is a high-precision tool designed for the calibration of measuring equipment or for gauging parts directly. Constructed from cold-stabilized tool steel, the gage offers durability and consistency for demanding inspection applications.
  • Diameter of 0.1257 in.
  • Overall length of 2 in.
  • Tolerance class X
  • Hardness of 60/65 Rc
  • Surface finish of 4 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards