Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 0.9950 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000511342
Encompass
This Class ZZ plug gage is designed for high-precision inspection and measurement applications. Crafted from tool steel with a 10 microfinish or better, it provides durable and accurate performance for tasks such as go inspection and the calibration of micrometers, verniers, and other measuring instruments. It conforms to ASME B89.1 standards.
  • Diameter of 0.9950 in.
  • Overall length of 2.0000 in.
  • Tolerance of +.0002/-.0000
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Designed for go inspection
Catalog No. 50-005-11342 Supplier MSC Supplier No. 88999503
This Class ZZ plug gage is designed for high-precision inspection and measurement applications. Crafted from tool steel with a 10 microfinish or better, it provides durable and accurate performance for tasks such as go inspection and the calibration of micrometers, verniers, and other measuring instruments. It conforms to ASME B89.1 standards.
  • Diameter of 0.9950 in.
  • Overall length of 2.0000 in.
  • Tolerance of +.0002/-.0000
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Designed for go inspection